Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAGGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ixodes scapularis | isc-miR-315 | MIMAT0012698 |
Stem-loop Details
Ixodes scapularis

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | isc-miR-315 |
---|---|
miRBase Accession Number: | MI0012278 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAGGC |
Species: | Ixodes scapularis |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAGGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ixodes scapularis | isc-miR-315 | MIMAT0012698 |