Assay Details
Gene Family ID: | MIPF0000821, mir-2241 |
Mature miRNA Sequence: | UUCGCAAGGAUUCCUCAAGAUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pristionchus pacificus | ppc-miR-2241a | MIMAT0011731 |
Stem-loop Details
Pristionchus pacificus

Genomic Map