Assay Details
Mature miRNA Sequence: | UCCCUGAGUUCAUGCCGUUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis remanei | crm-miR-2231-3p | MIMAT0011632 |
Stem-loop Details
Caenorhabditis remanei
More Information
Related Products
miRBase Alias:
Caenorhabditis remanei: crm-miR-2231 (v18)

Genomic Map