Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | ACUGUCUAACAACCAUCAAAUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schmidtea mediterranea | sme-miR-315-3p | MIMAT0012167 |
Stem-loop Details
Schmidtea mediterranea
More Information
Related Products
miRBase Alias:
Schmidtea mediterranea: sme-miR-315* (v18)

Genomic Map