Assay Details
Gene Family ID: | MIPF0000027, mir-23 |
Mature miRNA Sequence: | AUCACAUUGCCAGGGAUUACCAC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-23b-3p | MIMAT0000418 | |
| Alligator mississippiensis | ami-miR-23b-3p | MIMAT0038060 | |
| Bovine | bta-miR-23b-3p | MIMAT0003852 | |
| Dasypus novemcinctus | dno-miR-23b-3p | MIMAT0047531 | |
| Guinea pig | cpo-miR-23b-3p | MIMAT0046879 | |
| Ophiophagus hannah | oha-miR-23b-3p | MIMAT0036857 | |
| View More Rows | |||
| Oryzias latipes | ola-miR-23b | MIMAT0022588 | |
| Pan paniscus | ppa-miR-23b | MIMAT0002212 | |
| Pan troglodytes | ptr-miR-23b | MIMAT0002210 | |
| Petromyzon marinus | pma-miR-23b | MIMAT0019404 | |
| Pongo pygmaeus | ppy-miR-23b | MIMAT0002211 | |
| Python bivittatus | pbv-miR-23b-3p | MIMAT0038855 | |
| Rabbit | ocu-miR-23b-3p | MIMAT0048129 | |
| Zebrafish | dre-miR-23b-3p | MIMAT0048661 | |
Stem-loop Details
Human
Alligator mississippiensis
Bovine
Dasypus novemcinctus
Guinea pig
Ophiophagus hannah
Oryzias latipes
Pan paniscus
Pan troglodytes
Petromyzon marinus
Pongo pygmaeus
Python bivittatus
Rabbit
Zebrafish

Genomic Map