Assay Details
Gene Family ID: | MIPF0000065, mir-132 |
Mature miRNA Sequence: | ACCUUGGCUCUAGACUGCUUACU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-212-5p | MIMAT0022695 | |
| Mouse | mmu-miR-212-5p | MIMAT0017053 | |
| Bovine | bta-miR-212 | MIMAT0009264 | |
| Chicken | gga-miR-212-5p | MIMAT0049954 | |
| Cricetulus griseus | cgr-miR-212 | MIMAT0023850 | |
| Dog | cfa-miR-212 | MIMAT0006731 | |
| View More Rows | |||
| Guinea pig | cpo-miR-212-5p | MIMAT0047113 | |
| Ictalurus punctatus | ipu-miR-212 | MIMAT0029465 | |
| Monodelphis domestica | mdo-miR-212-5p | MIMAT0026692 | |
| Neolamprologus brichardi | nbr-miR-212b | MIMAT0042620 | |
| Pig | ssc-miR-212 | MIMAT0025370 | |
| Pteropus alecto | pal-miR-212-5p | MIMAT0039979 | |
| Python bivittatus | pbv-miR-212-5p | MIMAT0039016 | |
| Rabbit | ocu-miR-212-5p | MIMAT0048335 | |
| Rhesus monkey | mml-miR-212-5p | MIMAT0026847 | |
| Salmo salar | ssa-miR-212b-5p | MIMAT0032512 | |
| Zebrafish | dre-miR-212-5p | MIMAT0048650 | |
Stem-loop Details
Human
Mouse
Bovine
Chicken
Cricetulus griseus
Dog
Guinea pig
Ictalurus punctatus
Monodelphis domestica
Neolamprologus brichardi
Pig
Pteropus alecto
Python bivittatus
Rabbit
Rhesus monkey
Salmo salar
Zebrafish

Genomic Map