Assay Details
Gene Family ID: | MIPF0000056, mir-148 |
Mature miRNA Sequence: | GAAGUUCUGUUAUACACUCAGGCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-148b-5p | MIMAT0017036 |
Dasypus novemcinctus | dno-miR-148b-5p | MIMAT0047671 |
Guinea pig | cpo-miR-148b-5p | MIMAT0047024 |
Rabbit | ocu-miR-148b-5p | MIMAT0048261 |
Rhesus monkey | mml-miR-148b-5p | MIMAT0026826 |
Stem-loop Details
Mouse
Dasypus novemcinctus
Guinea pig
Rabbit
Rhesus monkey
