Assay Details
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | CAGCUAACAUGCAACUGCUCUC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-449a-3p | MIMAT0017181 |
Stem-loop Details
Rat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | rno-miR-449a* |
---|---|
miRBase Accession Number: | MI0001650 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CAGCUAACAUGCAACUGCUCUC |
Species: | Rat |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | CAGCUAACAUGCAACUGCUCUC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-449a-3p | MIMAT0017181 |