Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | CUUUCGAGCAAUAAUUGAAACC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-315-3p | MIMAT0020841 |
Stem-loop Details
Drosophila melanogaster

Genomic Map