Assay Details
Gene Family ID: | MIPF0000360, mir-551 |
Mature miRNA Sequence: | GAAAUCAAGCUUGGGUGAGACCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-551b-5p | MIMAT0017236 |
Rat | rno-miR-551b-5p | MIMAT0017335 |
Stem-loop Details
Mouse
Rat

More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-551b* (v17)
Rat: rno-miR-551b* (v18)
Rat: rno-miR-551b* (v18)
