Assay Details
Gene Family ID: | MIPF0000288, mir-374 |
Mature miRNA Sequence: | CUUAUCAGGUUGUAUUAUCAUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cricetulus griseus | cgr-miR-374-3p | MIMAT0023940 |
| Goat | chi-miR-374b-3p | MIMAT0036183 |
| Pig | ssc-miR-374b-3p | MIMAT0017378 |
Stem-loop Details
Cricetulus griseus
Goat
Pig

Genomic Map