Assay Details
Gene Family ID: | MIPF0000985, mir-1013 |
Mature miRNA Sequence: | AUAAAAGUAUGCCGAACUCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-1013-3p | MIMAT0005026 |
Stem-loop Details
Drosophila melanogaster

Genomic Map