Assay Details
Gene Family ID: | MIPF0000095, mir-122 |
Mature miRNA Sequence: | AAACACCAUUGUCACACUCCAC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-122b-3p | MIMAT0019877 |
| Mouse | mmu-miR-122b-3p | MIMAT0049853 |
Stem-loop Details
Human
Mouse

Genomic Map