Assay Details
Gene Family ID: | MIPF0001258, mir-4771 |
Mature miRNA Sequence: | AGCAGACUUGACCUACAAUUA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4771 | MIMAT0019925 |
Stem-loop Details
Human

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4771 |
---|---|
miRBase Accession Number: | MI0017412 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGCAGACUUGACCUACAAUUA |
Species: | Human |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0001258, mir-4771 |
Mature miRNA Sequence: | AGCAGACUUGACCUACAAUUA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4771 | MIMAT0019925 |