Assay Details
Gene Family ID: | MIPF0000024, mir-103 |
Mature miRNA Sequence: | UCGGCUUCUUUACAGUGCUGCCUUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-103-1-5p | MIMAT0031070 |
| Monodelphis domestica | mdo-miR-103-1-5p | MIMAT0026658 |
| Ornithorhynchus anatinus | oan-miR-103-1-5p | MIMAT0031098 |
| Pteropus alecto | pal-miR-103a-5p | MIMAT0040193 |
| Rhesus monkey | mml-miR-103-5p | MIMAT0026582 |
| Xenopus laevis | xla-miR-103a-5p | MIMAT0046414 |
Stem-loop Details
Chicken
Monodelphis domestica
Ornithorhynchus anatinus
Pteropus alecto
Rhesus monkey
Xenopus laevis

Genomic Map