Assay Details
Gene Family ID: | MIPF0000109, mir-186 |
Mature miRNA Sequence: | CCCAAAGGUGAAUUUUUUGGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Monodelphis domestica | mdo-miR-186-3p | MIMAT0026676 |
Stem-loop Details
Monodelphis domestica

Genomic Map