Assay Details
Mature miRNA Sequence: | GAGCUGCCCCUUGAAAGACUAA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Brugia malayi | bma-miR-1822 | MIMAT0031740 |
Stem-loop Details
Brugia malayi
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | bma-miR-1822 |
|---|---|
| miRBase Accession Number: | MI0026289 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAGCUGCCCCUUGAAAGACUAA |
| Species: | Brugia malayi |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | GAGCUGCCCCUUGAAAGACUAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Brugia malayi | bma-miR-1822 | MIMAT0031740 |