Assay Details
Gene Family ID: | MIPF0000293, mir-67 |
Mature miRNA Sequence: | UCACAACCUCCUUGAGUAAGCGGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Brugia malayi | bma-miR-307 | MIMAT0031748 |
Stem-loop Details
Brugia malayi
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | bma-miR-307 |
|---|---|
| miRBase Accession Number: | MI0026299 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCACAACCUCCUUGAGUAAGCGGA |
| Species: | Brugia malayi |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000293, mir-67 |
Mature miRNA Sequence: | UCACAACCUCCUUGAGUAAGCGGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Brugia malayi | bma-miR-307 | MIMAT0031748 |