Assay Details
Gene Family ID: | MIPF0000485, mir-1542 |
Mature miRNA Sequence: | CUGGCACAUAGGAGUUUCAUAAA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Monodelphis domestica | mdo-miR-1542-1-5p | MIMAT0026766 |
Stem-loop Details
Monodelphis domestica

Genomic Map