Assay Details
Gene Family ID: | MIPF0001700, mir-2068 |
Mature miRNA Sequence: | GGGCAUGUGUGCAUAGGGAAGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma belcheri | bbe-miR-2068-3p | MIMAT0031634 |
Stem-loop Details
Branchiostoma belcheri

Genomic Map