Assay Details
Gene Family ID: | MIPF0001681, mir-2071 |
Mature miRNA Sequence: | AUGCGGUGCGGUGGUAGCAACCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma belcheri | bbe-miR-2071-3p | MIMAT0031638 |
Stem-loop Details
Branchiostoma belcheri

Genomic Map