Assay Details
Gene Family ID: | MIPF0000203, mir-328 |
Mature miRNA Sequence: | CUGGCCCUCUCUGCCCUUCCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-328 | MIMAT0024166 |
| Rabbit | ocu-miR-328-3p | MIMAT0048377 |
Stem-loop Details
Gorilla gorilla
Rabbit

Genomic Map