Assay Details
Gene Family ID: | MIPF0000163, mir-320 |
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAAGGUGA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pan troglodytes | ptr-miR-320e | MIMAT0024030 |
Pongo pygmaeus | ppy-miR-320e | MIMAT0024358 |
Stem-loop Details
Pan troglodytes
Pongo pygmaeus
