Assay Details
Gene Family ID: | MIPF0000143, mir-326 |
Mature miRNA Sequence: | CCUCGUCUGUCUGUUGGGCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Goat | chi-miR-326-5p | MIMAT0036137 |
Stem-loop Details
Goat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | chi-miR-326-5p |
|---|---|
| miRBase Accession Number: | MI0030741 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUCGUCUGUCUGUUGGGCU |
| Species: | Goat |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000143, mir-326 |
Mature miRNA Sequence: | CCUCGUCUGUCUGUUGGGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Goat | chi-miR-326-5p | MIMAT0036137 |