Assay Details
Gene Family ID: | MIPF0000825, mir-2235 |
Mature miRNA Sequence: | UCACCGGAAGCAUUGUAUGAUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pristionchus pacificus | ppc-miR-2235a-10-3p | MIMAT0033005 |
Stem-loop Details
Pristionchus pacificus

Genomic Map