Assay Details
Mature miRNA Sequence: | GUAACGGAACCCAUAAUGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Oryzias latipes | ola-miR-462 | MIMAT0022561 |
Stem-loop Details
Oryzias latipes
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ola-miR-462 |
|---|---|
| miRBase Accession Number: | MI0019439 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUAACGGAACCCAUAAUGC |
| Species: | Oryzias latipes |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | GUAACGGAACCCAUAAUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Oryzias latipes | ola-miR-462 | MIMAT0022561 |