Assay Details
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | AGGACGGUAUUUGUGACAAUAUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila virilis | dvi-miR-311-5p | MIMAT0035566 |
Stem-loop Details
Drosophila virilis
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dvi-miR-311-5p |
|---|---|
| miRBase Accession Number: | MI0029246 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGACGGUAUUUGUGACAAUAUG |
| Species: | Drosophila virilis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | AGGACGGUAUUUGUGACAAUAUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila virilis | dvi-miR-311-5p | MIMAT0035566 |