Assay Details
Gene Family ID: | MIPF0000138, mir-363 |
Mature miRNA Sequence: | UGGCUGGUUUGCUGUUGUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Sarcophilus harrisii | sha-miR-363 | MIMAT0022831 |
Stem-loop Details
Sarcophilus harrisii
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | sha-miR-363 |
|---|---|
| miRBase Accession Number: | MI0019660 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGCUGGUUUGCUGUUGUC |
| Species: | Sarcophilus harrisii |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000138, mir-363 |
Mature miRNA Sequence: | UGGCUGGUUUGCUGUUGUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Sarcophilus harrisii | sha-miR-363 | MIMAT0022831 |