Assay Details
Gene Family ID: | MIPF0000174, mir-454 |
Mature miRNA Sequence: | UAGUGCAAUAUUGCUUAUAGGGUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-454 | MIMAT0024280 |
Stem-loop Details
Gorilla gorilla
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ggo-miR-454 |
|---|---|
| miRBase Accession Number: | MI0020828 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGUGCAAUAUUGCUUAUAGGGUU |
| Species: | Gorilla gorilla |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000174, mir-454 |
Mature miRNA Sequence: | UAGUGCAAUAUUGCUUAUAGGGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-454 | MIMAT0024280 |