Assay Details
Gene Family ID: | MIPF0001399, mir-2110 |
Mature miRNA Sequence: | UUGGGGAAGCGGCCGCUGAGUGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-2110 | MIMAT0024250 |
Stem-loop Details
Gorilla gorilla
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ggo-miR-2110 |
|---|---|
| miRBase Accession Number: | MI0020798 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGGGGAAGCGGCCGCUGAGUGA |
| Species: | Gorilla gorilla |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0001399, mir-2110 |
Mature miRNA Sequence: | UUGGGGAAGCGGCCGCUGAGUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-2110 | MIMAT0024250 |