Assay Details
Gene Family ID: | MIPF0000558, mir-1307 |
Mature miRNA Sequence: | UCGACCGGACCUCGACCGGCUCGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Eptesicus fuscus | efu-miR-1307 | MIMAT0034967 |
Stem-loop Details
Eptesicus fuscus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | efu-miR-1307 |
|---|---|
| miRBase Accession Number: | MI0028652 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCGACCGGACCUCGACCGGCUCGU |
| Species: | Eptesicus fuscus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000558, mir-1307 |
Mature miRNA Sequence: | UCGACCGGACCUCGACCGGCUCGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Eptesicus fuscus | efu-miR-1307 | MIMAT0034967 |