Assay Details
Gene Family ID: | MIPF0000610, mir-1290 |
Mature miRNA Sequence: | UGGAUUUUUGGAUCAGGGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1290 | MIMAT0005880 |
| Pan troglodytes | ptr-miR-1290 | MIMAT0008008 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map