Assay Details
Gene Family ID: | MIPF0000027, mir-23 |
Mature miRNA Sequence: | UGGGUUCCUGGCAUGCUGAUUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-23b-5p | MIMAT0004587 |
| Bovine | bta-miR-10174-5p | MIMAT0040927 |
| Goat | chi-miR-23b-5p | MIMAT0036088 |
| Ophiophagus hannah | oha-miR-23b-5p | MIMAT0036856 |
| Ornithorhynchus anatinus | oan-miR-23b-5p | MIMAT0006958 |
Stem-loop Details
Human
Bovine
Goat
Ophiophagus hannah
Ornithorhynchus anatinus

Genomic Map