Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUAUAACACAGAUGGCCUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-410-3p | MIMAT0002171 |
| Mouse | mmu-miR-410-3p | MIMAT0001091 |
| Rat | rno-miR-410-3p | MIMAT0005311 |
| Bovine | bta-miR-410 | MIMAT0009311 |
| Callithrix jacchus | cja-miR-410 | MIMAT0039354 |
| Cricetulus griseus | cgr-miR-410-3p | MIMAT0023953 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-410-3p | MIMAT0047873 |
| Dog | cfa-miR-410 | MIMAT0006728 |
| Goat | chi-miR-410-3p | MIMAT0036212 |
| Guinea pig | cpo-miR-410-3p | MIMAT0047236 |
| Horse | eca-miR-410 | MIMAT0013153 |
| Microcebus murinus | mmr-miR-410 | MIMAT0049189 |
| Nomascus leucogenys | nle-miR-410 | MIMAT0049369 |
| Otolemur garnettii | oga-miR-410 | MIMAT0049673 |
| Pan paniscus | ppa-miR-410 | MIMAT0049033 |
| Pan troglodytes | ptr-miR-410 | MIMAT0008137 |
| Pongo pygmaeus | ppy-miR-410 | MIMAT0015877 |
| Pteropus alecto | pal-miR-410-3p | MIMAT0040152 |
| Rabbit | ocu-miR-410-3p | MIMAT0048436 |
| Rhesus monkey | mml-miR-410-3p | MIMAT0006316 |
| Sheep | oar-miR-410-3p | MIMAT0019334 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-410 |
| Stem-loop Accession # | MI0002465 |
| Stem-loop Sequence | GGUACCUGAGAAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC |
| Chromosome Location | Chr. 14 - 101065912 - 101065991 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002171
miRBase ID: hsa-miR-410-3p Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: Chr. 14 - 101065912 - 101065991 [+] on Build GRCh38 |
| Stem-loop ID | mmu-mir-410 |
| Stem-loop Accession # | MI0001161 |
| Stem-loop Sequence | GGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACCA |
| Chromosome Location | Chr. 12 - 109743715 - 109743795 [+] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001091
miRBase ID: mmu-miR-410-3p Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: Chr. 12 - 109743715 - 109743795 [+] on Build GRCm38 |
| Stem-loop ID | rno-mir-410 |
| Stem-loop Accession # | MI0006143 |
| Stem-loop Sequence | ACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACC |
| Chromosome Location | Chr. 6 - 133893997 - 133894072 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005311
miRBase ID: rno-miR-410-3p Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: Chr. 6 - 133893997 - 133894072 [+] on Build Rnor_6.0 |
| Stem-loop ID | bta-mir-410 |
| Stem-loop Accession # | MI0009825 |
| Stem-loop Sequence | GGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009311
miRBase ID: bta-miR-410 Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | cja-mir-410 |
| Stem-loop Accession # | MI0031868 |
| Stem-loop Sequence | GGUACCUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039354
miRBase ID: cja-miR-410 Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | cgr-mir-410 |
| Stem-loop Accession # | MI0020521 |
| Stem-loop Sequence | CUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0023953
miRBase ID: cgr-miR-410-3p Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | dno-mir-410 |
| Stem-loop Accession # | MI0039101 |
| Stem-loop Sequence | AGGCUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047873
miRBase ID: dno-miR-410-3p Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | cfa-mir-410 |
| Stem-loop Accession # | MI0008149 |
| Stem-loop Sequence | AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGCCGAAUAUAACACAGAUGGCCUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006728
miRBase ID: cfa-miR-410 Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | chi-mir-410 |
| Stem-loop Accession # | MI0030783 |
| Stem-loop Sequence | CACCUGCGGUCUGUGGAGUAGGCGCCACUUUGGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACCACCCACCCCUCGUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036212
miRBase ID: chi-miR-410-3p Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | cpo-mir-410 |
| Stem-loop Accession # | MI0038766 |
| Stem-loop Sequence | AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047236
miRBase ID: cpo-miR-410-3p Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | eca-mir-410 |
| Stem-loop Accession # | MI0012897 |
| Stem-loop Sequence | UGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUCUUCAGUACCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013153
miRBase ID: eca-miR-410 Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | mmr-mir-410 |
| Stem-loop Accession # | MI0039908 |
| Stem-loop Sequence | AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049189
miRBase ID: mmr-miR-410 Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | nle-mir-410 |
| Stem-loop Accession # | MI0040106 |
| Stem-loop Sequence | AGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049369
miRBase ID: nle-miR-410 Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | oga-mir-410 |
| Stem-loop Accession # | MI0040456 |
| Stem-loop Sequence | AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049673
miRBase ID: oga-miR-410 Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | ppa-mir-410 |
| Stem-loop Accession # | MI0039741 |
| Stem-loop Sequence | AGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049033
miRBase ID: ppa-miR-410 Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | ptr-mir-410 |
| Stem-loop Accession # | MI0008658 |
| Stem-loop Sequence | GUACCUGAGAAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008137
miRBase ID: ptr-miR-410 Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | ppy-mir-410 |
| Stem-loop Accession # | MI0014934 |
| Stem-loop Sequence | GGUACCUGAGAAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015877
miRBase ID: ppy-miR-410 Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | pal-mir-410 |
| Stem-loop Accession # | MI0032520 |
| Stem-loop Sequence | AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0040152
miRBase ID: pal-miR-410-3p Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | ocu-mir-410 |
| Stem-loop Accession # | MI0039399 |
| Stem-loop Sequence | AGGUUGUCUGUGAUGAGUUCGCUGUAUUAAAGCCGAAUAUAACACAGAUGGCCUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048436
miRBase ID: ocu-miR-410-3p Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | mml-mir-410 |
| Stem-loop Accession # | MI0007734 |
| Stem-loop Sequence | GGUACCUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006316
miRBase ID: mml-miR-410-3p Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
| Stem-loop ID | oar-mir-410 |
| Stem-loop Accession # | MI0016961 |
| Stem-loop Sequence | CACCUGCGGUCUGUGGAGUAGGCGCCACUUUGGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACCACCCACCCCUCGUGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0019334
miRBase ID: oar-miR-410-3p Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU Chromosome Location: NA |
More Information
Related Products

Genomic Map