Assay Details
Gene Family ID: | MIPF0000063, mir-192 |
Mature miRNA Sequence: | AUGACCUAUGAAUUGACAGAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-215-5p | MIMAT0000272 |
| Alligator mississippiensis | ami-miR-215-5p | MIMAT0038240 |
| Chicken | gga-miR-215-5p | MIMAT0001134 |
| Chrysemys picta | cpi-miR-215-5p | MIMAT0037841 |
| Goat | chi-miR-215-5p | MIMAT0036060 |
| Gorilla gorilla | ggo-miR-215 | MIMAT0002734 |
| View More Rows | ||
| Horse | eca-miR-215 | MIMAT0013190 |
| Macaca nemestrina | mne-miR-215 | MIMAT0002736 |
| Ornithorhynchus anatinus | oan-miR-215-5p | MIMAT0007000 |
| Pan troglodytes | ptr-miR-215 | MIMAT0002730 |
| Pig | ssc-miR-215 | MIMAT0010192 |
| Pongo pygmaeus | ppy-miR-215 | MIMAT0002732 |
| Rhesus monkey | mml-miR-215-5p | MIMAT0002728 |
| Taeniopygia guttata | tgu-miR-215-5p | MIMAT0014577 |
Stem-loop Details
Human
Alligator mississippiensis
Chicken
Chrysemys picta
Goat
Gorilla gorilla
Horse
Macaca nemestrina
Ornithorhynchus anatinus
Pan troglodytes
Pig
Pongo pygmaeus
Rhesus monkey
Taeniopygia guttata

Genomic Map