Assay Details
Gene Family ID: | MIPF0000163, mir-320 |
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGGCAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-320b | MIMAT0005792 |
| Pan troglodytes | ptr-miR-320b | MIMAT0008094 |
| Pongo pygmaeus | ppy-miR-320b | MIMAT0015822 |
| Rhesus monkey | mml-miR-320b | MIMAT0024312 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map