Assay Details
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | UUUUGCACCUUUUGGAGUGAA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-507 | MIMAT0002879 |
Pan troglodytes | ptr-miR-507 | MIMAT0005771 |
Pongo pygmaeus | ppy-miR-507 | MIMAT0015930 |
Pygathrix bieti | pbi-miR-507 | MIMAT0005780 |
Rhesus monkey | mml-miR-507 | MIMAT0005763 |
Symphalangus syndactylus | ssy-miR-507 | MIMAT0005753 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Pygathrix bieti
Rhesus monkey
Symphalangus syndactylus
