Assay Details
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | GUGUCUUUUGCUCUGCAGUCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-511-5p | MIMAT0002808 |
| Pan troglodytes | ptr-miR-511 | MIMAT0008179 |
| Pongo pygmaeus | ppy-miR-511 | MIMAT0015936 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus

Genomic Map