Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AUCGUGCAUCCUUUUAGAGUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-517c-3p | MIMAT0002866 |
| Pongo pygmaeus | ppy-miR-517c-?? | MIMAT0036449 |
| Pongo pygmaeus | ppy-miR-517c-3p | MIMAT0036448 |
| Rhesus monkey | mml-miR-517a | MIMAT0006375 |
Stem-loop Details
Human
Pongo pygmaeus
Rhesus monkey

Genomic Map