Assay Details
Gene Family ID: | MIPF0001805, mir-6505 |
Mature miRNA Sequence: | UUGGAAUAGGGGAUAUCUCAGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-6505-5p | MIMAT0025466 |
| Rhesus monkey | mml-miR-6505-5p | MIMAT0028248 |
Stem-loop Details
Human
Rhesus monkey

Genomic Map