Assay Details
Gene Family ID: | MIPF0000019, mir-8 |
Mature miRNA Sequence: | CAUCUUACCGGACAGUGCUGGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-200a-5p | MIMAT0001620 |
Mouse | mmu-miR-200a-5p | MIMAT0004619 |
Dog | cfa-miR-200a | MIMAT0009865 |
Guinea pig | cpo-miR-200a-5p | MIMAT0047089 |
Pteropus alecto | pal-miR-200a-5p | MIMAT0039866 |
Rhesus monkey | mml-miR-200a-5p | MIMAT0026842 |
View More Rows | ||
Zebrafish | dre-miR-200a-5p | MIMAT0031984 |
Stem-loop Details
Human
Mouse
Dog
Guinea pig
Pteropus alecto
Rhesus monkey
Zebrafish

More Information
Related Products
miRBase Alias:
Human: hsa-miR-200a* (v17)
Mouse: mmu-miR-200a* (v17)
Mouse: mmu-miR-200a* (v17)
