Assay Details
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | UGUAAACAUCCUACACUCAGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30b-5p | MIMAT0000420 |
| Mouse | mmu-miR-30b-5p | MIMAT0000130 |
| Rat | rno-miR-30b-5p | MIMAT0000806 |
| Alligator mississippiensis | ami-miR-30b-5p | MIMAT0038079 |
| Anolis carolinensis | aca-miR-30b-5p | MIMAT0021922 |
| Astatotilapia burtoni | abu-miR-30d | MIMAT0042084 |
| View More Rows | ||
| Bovine | bta-miR-30b-5p | MIMAT0003547 |
| Callithrix jacchus | cja-miR-30b | MIMAT0039502 |
| Chicken | gga-miR-30b-5p | MIMAT0001130 |
| Chrysemys picta | cpi-miR-30b-5p | MIMAT0037676 |
| Cricetulus griseus | cgr-miR-30b-5p | MIMAT0023896 |
| Cyprinus carpio | ccr-miR-30b | MIMAT0026289 |
| Dasypus novemcinctus | dno-miR-30b-5p | MIMAT0047558 |
| Dog | cfa-miR-30b | MIMAT0006617 |
| Fugu rubripes | fru-miR-30b | MIMAT0002947 |
| Gadus morhua | gmo-miR-30c-5p | MIMAT0044142 |
| Goat | chi-miR-30b-5p | MIMAT0036123 |
| Guinea pig | cpo-miR-30b-5p | MIMAT0046905 |
| Horse | eca-miR-30b | MIMAT0013005 |
| Ictalurus punctatus | ipu-miR-30b | MIMAT0029497 |
| Metriaclima zebra | mze-miR-30d | MIMAT0042339 |
| Monodelphis domestica | mdo-miR-30b-5p | MIMAT0028550 |
| Neolamprologus brichardi | nbr-miR-30d | MIMAT0042527 |
| Nomascus leucogenys | nle-miR-30b | MIMAT0049368 |
| Ophiophagus hannah | oha-miR-30b-5p | MIMAT0036890 |
| Oreochromis niloticus | oni-miR-30d | MIMAT0042707 |
| Ornithorhynchus anatinus | oan-miR-30b-5p | MIMAT0007043 |
| Otolemur garnettii | oga-miR-30b | MIMAT0049616 |
| Pig | ssc-miR-30b-5p | MIMAT0007756 |
| Pongo pygmaeus | ppy-miR-30b | MIMAT0015736 |
| Pteropus alecto | pal-miR-30b-5p | MIMAT0039945 |
| Pundamilia nyererei | pny-miR-30d | MIMAT0042905 |
| Python bivittatus | pbv-miR-30b-5p | MIMAT0038872 |
| Rabbit | ocu-miR-30b-5p | MIMAT0048152 |
| Salmo salar | ssa-miR-30e-5p | MIMAT0032611 |
| Taeniopygia guttata | tgu-miR-30e | MIMAT0025411 |
| Tetraodon nigroviridis | tni-miR-30b | MIMAT0002948 |
| Tupaia chinensis | tch-miR-30b-5p | MIMAT0036503 |
| Xenopus laevis | xla-miR-30b-5p | MIMAT0046570 |
| Xenopus tropicalis | xtr-miR-30b | MIMAT0003576 |
| Zebrafish | dre-miR-30b | MIMAT0001804 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-30b |
| Stem-loop Accession # | MI0000441 |
| Stem-loop Sequence | ACCAAGUUUCAGUUCAUGUAAACAUCCUACACUCAGCUGUAAUACAUGGAUUGGCUGGGAGGUGGAUGUUUACUUCAGCUGACUUGGA |
| Chromosome Location | Chr. 8 - 134800520 - 134800607 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000420
miRBase ID: hsa-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: Chr. 8 - 134800520 - 134800607 [-] on Build GRCh38 |
| Stem-loop ID | mmu-mir-30b |
| Stem-loop Accession # | MI0000145 |
| Stem-loop Sequence | CUAAGCCAAGUUUCAGUUCAUGUAAACAUCCUACACUCAGCUGUCAUACAUGCGUUGGCUGGGAUGUGGAUGUUUACGUCAGCUGUCUUGGAGUAU |
| Chromosome Location | Chr. 15 - 68337415 - 68337510 [-] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000130
miRBase ID: mmu-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: Chr. 15 - 68337415 - 68337510 [-] on Build GRCm38 |
| Stem-loop ID | rno-mir-30b |
| Stem-loop Accession # | MI0000868 |
| Stem-loop Sequence | CCGAGUUUCAGUUCAUGUAAACAUCCUACACUCAGCUGUCAUACAUGAGUUGGCUGGGAUGUGGAUGUUUACGUCAGCUGUCUUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000806
miRBase ID: rno-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | ami-mir-30b |
| Stem-loop Accession # | MI0029691 |
| Stem-loop Sequence | ACUGACUUUUAGUUCAUGUAAACAUCCUACACUCAGCUAUAACAACUGGAACGGCUGGGAGGUGGAUGUUUACUUCAACUGAUUUGAAAAGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038079
miRBase ID: ami-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | aca-mir-30b |
| Stem-loop Accession # | MI0018848 |
| Stem-loop Sequence | UACUGAUUUUUAGUUCAUGUAAACAUCCUACACUCAGCUGUAUCACAUGAAAUGGCUGGGAGAUGAAUGUUUACUUCAACUGAUUUGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0021922
miRBase ID: aca-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | abu-mir-30d |
| Stem-loop Accession # | MI0033781 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUUAUUGAUUGCCACAGAGGCUGAGAGGAGGUUGUUUACUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042084
miRBase ID: abu-miR-30d Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | bta-mir-30b |
| Stem-loop Accession # | MI0004761 |
| Stem-loop Sequence | CCAAGUUUUCAGUUCAUGUAAACAUCCUACACUCAGCUGUAACACACGAGUCGGCUGGGAGGUGGAUGUUUACUUCAGCUGACUUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003547
miRBase ID: bta-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | cja-mir-30b |
| Stem-loop Accession # | MI0032028 |
| Stem-loop Sequence | CCGAGUUUCAGUUCAUGUAAACAUCCUACACUCAGCUGUAAUACAUGGAUUGGCUGAGAGGUGGAUGUUUACUUCAGCUGACUUGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039502
miRBase ID: cja-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | gga-mir-30b |
| Stem-loop Accession # | MI0001199 |
| Stem-loop Sequence | CUAACUUUUAGUUCCUGUAAACAUCCUACACUCAGCUAUAACAAGUGGUAGGGCUGGGGGGUGGAUGUUUACUUCAACUGACUUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001130
miRBase ID: gga-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | cpi-mir-30b |
| Stem-loop Accession # | MI0029425 |
| Stem-loop Sequence | UACUGAUUUUUAGUUCAUGUAAACAUCCUACACUCAGCUAUAAAACAUAGAACGGCUGGGAGGUGGAUGUUUACUUCAACUGACUUGGAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037676
miRBase ID: cpi-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | cgr-mir-30b |
| Stem-loop Accession # | MI0020485 |
| Stem-loop Sequence | GUUCAGUUCAUGUAAACAUCCUACACUCAGCUGUCAUACAUGAGUUGGCUGGGAUGUGGAUGUUUACGUCAGCUGUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0023896
miRBase ID: cgr-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | ccr-mir-30b |
| Stem-loop Accession # | MI0023389 |
| Stem-loop Sequence | UAAUAUUCCAGUGUAGUCGUUGUAAACAUCCUACACUCAGCUGUAAUCUGCACAUGAGGCUGGGAGGAGGGUGUUUGCUGUGACUGACUGGAGCUCCUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0026289
miRBase ID: ccr-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | dno-mir-30b |
| Stem-loop Accession # | MI0038935 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUGAUGUGUGGAUUGGCUGGGAGGUGGAUGUUUACUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047558
miRBase ID: dno-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | cfa-mir-30b |
| Stem-loop Accession # | MI0008013 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUAGUACAUGGAUUGGCUGGGAGGUGGAUGUUUACUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006617
miRBase ID: cfa-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | fru-mir-30b |
| Stem-loop Accession # | MI0003263 |
| Stem-loop Sequence | UUCCGGCUCAGUUGGUGUAAACAUCCUACACUCAGCUGUGAUCCUUUGCGGCAGAGGCUGAGAGAAGGUUGUUUACUUGAACUGGCUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002947
miRBase ID: fru-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-30c |
| Stem-loop Accession # | MI0036071 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUAGCCACGUUGACCACAGGCUGAGAGAAGGCUGUUUACUUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044142
miRBase ID: gmo-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | chi-mir-30b |
| Stem-loop Accession # | MI0030732 |
| Stem-loop Sequence | ACCAAGUUUCCAGUUCAUGUAAACAUCCUACACUCAGCUGUAACACUUGAGUCGGCUGGGAGGUGGAUGUUUACUUCAGCUGACUUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036123
miRBase ID: chi-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | cpo-mir-30b |
| Stem-loop Accession # | MI0038591 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUAAUAUAUGAAUUGGCUGGGAGGUGGGUGUUUACUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046905
miRBase ID: cpo-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | eca-mir-30b |
| Stem-loop Accession # | MI0012757 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUACUACAUGGAUUGGCUGGGAGGUGGAUGUUUACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013005
miRBase ID: eca-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | ipu-mir-30b |
| Stem-loop Accession # | MI0024614 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUGAGCUGCAGACGAGGCUGGGCGGAGGGUGUUUGCUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0029497
miRBase ID: ipu-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | mze-mir-30d |
| Stem-loop Accession # | MI0034110 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUUAUUGAUUGCCACAGAGGCUGAGAGGAGGUUGUUUACUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042339
miRBase ID: mze-miR-30d Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | mdo-mir-30b |
| Stem-loop Accession # | MI0023794 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUAACACAUGGAUCGGCUGGGAGGUGGAUGUUUACUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0028550
miRBase ID: mdo-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | nbr-mir-30d |
| Stem-loop Accession # | MI0034370 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUUAUUGAUUGCCACAGAGGCUGAGAGGAGGUUGUUUACUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042527
miRBase ID: nbr-miR-30d Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | nle-mir-30b |
| Stem-loop Accession # | MI0040105 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUAAUACAUGAAUUGGCUGGGAGGUGGAUGUUUGCUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049368
miRBase ID: nle-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | oha-mir-30b |
| Stem-loop Accession # | MI0031451 |
| Stem-loop Sequence | UGUUAGUUCGUGUAAACAUCCUACACUCAGCUUUUAUCCCAUGAAAUGGCUGGGGGAUGGAAGUUCACUUCAACUGAUUUGGAAAGAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036890
miRBase ID: oha-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | oni-mir-30d |
| Stem-loop Accession # | MI0034620 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUUAUUGAUUGCCACAGAGGCUGAGAGGAGGUUGUUUACUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042707
miRBase ID: oni-miR-30d Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | oan-mir-30b |
| Stem-loop Accession # | MI0006813 |
| Stem-loop Sequence | CCGGCCGGCAAAACCGAGUUCGAGUUCAUGUAAACAUCCUACACUCAGCUGUGACCCACCCAGCGGCUGGGAGGGGGAUGUUUGCUUCAACCGACUUGGAACCGCAGCCGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0007043
miRBase ID: oan-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | oga-mir-30b |
| Stem-loop Accession # | MI0040397 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUAAUAUAUGAAUUGGCUGGGAGGUGGAUGUUUACUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049616
miRBase ID: oga-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | ssc-mir-30b |
| Stem-loop Accession # | MI0008215 |
| Stem-loop Sequence | UUUCAGUUCGUGUAAACAUCCUACACUCAGCUGUAAUACAUGGAUUGGCUGGGAGGUGGAUGUUUACUUCAGCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0007756
miRBase ID: ssc-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | ppy-mir-30b |
| Stem-loop Accession # | MI0014802 |
| Stem-loop Sequence | ACCAAGUUUCAGUUCAUGUAAACAUCCUACACUCAGCUGUAAUACAUGGAUUGGCUGGGAGGUGGAUGUUUACUUCAGCUGACUUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015736
miRBase ID: ppy-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | pal-mir-30b |
| Stem-loop Accession # | MI0032413 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUAAUACAUGGAUUGGCUGGGAGGUGGAUGUUUACUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039945
miRBase ID: pal-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | pny-mir-30d |
| Stem-loop Accession # | MI0034879 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUUAUUGAUUGCCACAGAGGCUGAGAGGAGGUUGUUUACUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042905
miRBase ID: pny-miR-30d Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | pbv-mir-30b |
| Stem-loop Accession # | MI0030184 |
| Stem-loop Sequence | ACUGAUUCUUAGUUUAUGUAAACAUCCUACACUCAGCUUUAUCACAUGAAAUGGCUGGGAGAUGGAAGUUUACUUCAACUGAUUUGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038872
miRBase ID: pbv-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | ocu-mir-30b |
| Stem-loop Accession # | MI0039248 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUAAUACAUGAAUUGGCUGGGAGGUGGAUGUUUACGUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048152
miRBase ID: ocu-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | ssa-mir-30e-1 |
| Stem-loop Accession # | MI0026683 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUGAGUUGGCAGCAGAGGCUGGGAGAAGGCUGUUUACUUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032611
miRBase ID: ssa-miR-30e-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | ssa-mir-30e-2 |
| Stem-loop Accession # | MI0026684 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUGAGUUGGAAGCAGAGGCUGGGAGAAGGCUGUUUACUUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032611
miRBase ID: ssa-miR-30e-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | ssa-mir-30e-3 |
| Stem-loop Accession # | MI0026685 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUAUAGCCUGUAUGGAAGGCUGGGGAGAGGGUGUUUACUUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032611
miRBase ID: ssa-miR-30e-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | tgu-mir-30e |
| Stem-loop Accession # | MI0022188 |
| Stem-loop Sequence | UUUUAGUUCCUGUAAACAUCCUACACUCAGCUAUUAUAAGUGGUGGGGCUGGGGGUUGGGUGUUUACUUCAACUGACUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0025411
miRBase ID: tgu-miR-30e Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | tni-mir-30b |
| Stem-loop Accession # | MI0003264 |
| Stem-loop Sequence | AGAUGGUGUAAACAUCCUACACUCAGCUGUGAUCUUUUGCCGCAGAGGCUGAGAGAAGGUUGUUUACUUGAACUGGCUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002948
miRBase ID: tni-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | tch-mir-30b |
| Stem-loop Accession # | MI0031160 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUGUAAUGCAUGGAUUGGCUGGGAGGUGGAUGUUUACCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036503
miRBase ID: tch-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | xla-mir-30b |
| Stem-loop Accession # | MI0038318 |
| Stem-loop Sequence | UGUAAACAUCCUACACUCAGCUCGAAUGUAUAACCUGACUGGGUGGGGGGUGUUUGCCUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046570
miRBase ID: xla-miR-30b-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | xtr-mir-30b |
| Stem-loop Accession # | MI0004814 |
| Stem-loop Sequence | GUAUUAGUCUAUGUAAACAUCCUACACUCAGCUCGUAUGUAUAACCUGACUGGGUGGGGGGUGUUUGCCUCGACUGAUCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003576
miRBase ID: xtr-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
| Stem-loop ID | dre-mir-30b |
| Stem-loop Accession # | MI0001941 |
| Stem-loop Sequence | UUCCAGUGUAGUCGCUGUAAACAUCCUACACUCAGCUGUGAGCUGCAGACGAGGCUGGGCGGAGGGUGUUUGCUGUGACUGUCUGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001804
miRBase ID: dre-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCAGCU Chromosome Location: NA |
More Information
Related Products
miRBase Alias:
Human: hsa-miR-30b (v17)
Mouse: mmu-miR-30b (v17)
Rat: rno-miR-30b (v9.2)
Aeromonas caviae Ae398: aca-miR-30b (v18)
Bos taurus: bta-miR-30b (v13)
Gallus gallus: gga-miR-30b (v19)
Ochrobactrum anthropi: oan-miR-30b (v18)
Staphylococcus schleiferi: ssc-miR-30b (v14)
Mouse: mmu-miR-30b (v17)
Rat: rno-miR-30b (v9.2)
Aeromonas caviae Ae398: aca-miR-30b (v18)
Bos taurus: bta-miR-30b (v13)
Gallus gallus: gga-miR-30b (v19)
Ochrobactrum anthropi: oan-miR-30b (v18)
Staphylococcus schleiferi: ssc-miR-30b (v14)

Genomic Map