Assay Details
Mature miRNA Sequence: | AACAUUCAUUGUUGUCGGUGGGUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dasypus novemcinctus | dno-miR-181d-5p | MIMAT0047697 |
Eptesicus fuscus | efu-miR-181d | MIMAT0035133 |
Pan paniscus | ppa-miR-181d | MIMAT0049083 |
Pig | ssc-miR-181d-5p | MIMAT0013888 |
Pteropus alecto | pal-miR-181d-5p | MIMAT0040142 |
Rabbit | ocu-miR-181d-5p | MIMAT0048283 |
Stem-loop Details
Dasypus novemcinctus
Eptesicus fuscus
Pan paniscus
Pig
Pteropus alecto
Rabbit
