Assay Details
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | AGGCAGUGUAUUGUUAGCUGGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-449b-5p | MIMAT0003327 |
| Bovine | bta-miR-449b | MIMAT0009321 |
| Goat | chi-miR-449b-5p | MIMAT0036232 |
| Pan troglodytes | ptr-miR-449b | MIMAT0008150 |
| Pongo pygmaeus | ppy-miR-449b | MIMAT0015892 |
| Rhesus monkey | mml-miR-449b-5p | MIMAT0006331 |
Stem-loop Details
Human
Bovine
Goat
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-449b (v17)
Macaca mulatta: mml-miR-449b (v19)
Macaca mulatta: mml-miR-449b (v19)

Genomic Map