Assay Details
Gene Family ID: | MIPF0000395, mir-760 |
Mature miRNA Sequence: | CCCCUCAGGCCACCAGAGCCCG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-760-5p | MIMAT0005336 |
Stem-loop Details
Rat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | rno-miR-760-5p |
---|---|
miRBase Accession Number: | MI0006164 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CCCCUCAGGCCACCAGAGCCCG |
Species: | Rat |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000395, mir-760 |
Mature miRNA Sequence: | CCCCUCAGGCCACCAGAGCCCG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-760-5p | MIMAT0005336 |