Assay Details
Mature miRNA Sequence: | AGGUUGCCUCAUAGUGAGCUUGCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-453 | MIMAT0004870 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-453 |
|---|---|
| miRBase Accession Number: | MI0005497 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGUUGCCUCAUAGUGAGCUUGCA |
| Species: | Mouse |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | AGGUUGCCUCAUAGUGAGCUUGCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-453 | MIMAT0004870 |