Assay Details
Gene Family ID: | MIPF0002093, mir-3552 |
Mature miRNA Sequence: | AGGCUGCAGGCCCACUUCCCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-3552 | MIMAT0035715 |
| Rat | rno-miR-3552 | MIMAT0017813 |
Stem-loop Details
Mouse
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-3552 |
|---|---|
| miRBase Accession Number: | MI0030344 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGCUGCAGGCCCACUUCCCU |
| Species: | Mouse, Rat |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0002093, mir-3552 |
Mature miRNA Sequence: | AGGCUGCAGGCCCACUUCCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-3552 | MIMAT0035715 |
| Rat | rno-miR-3552 | MIMAT0017813 |