Assay Details
| Gene Family ID: | MIPF0000048, mir-128 | 
| Mature miRNA Sequence: | GGGGGCCGAUGCACUGUAAGAGA | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-128-2-5p | MIMAT0017069 | 
Stem-loop Details
Mouse
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-128-2-5p | 
|---|---|
| miRBase Accession Number: | MI0000726 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | GGGGGCCGAUGCACUGUAAGAGA | 
| Species: | Mouse | 
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor | 
| Gene Family ID: | MIPF0000048, mir-128 | 
| Mature miRNA Sequence: | GGGGGCCGAUGCACUGUAAGAGA | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-128-2-5p | MIMAT0017069 | 
 Back To Top
 Back To Top