Assay Details
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | UAGAACUUCGUCCCAAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-449c-3p | MIMAT0017804 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-449c-3p |
|---|---|
| miRBase Accession Number: | MI0015402 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGAACUUCGUCCCAAC |
| Species: | Rat |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | UAGAACUUCGUCCCAAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-449c-3p | MIMAT0017804 |