Assay Details
Amplicon Length: | 92 |
Chromosome Location: | Chr. 9 - 94175759 - 94175759 [-] on 94175759 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-let-7a-3p | MIMAT0004481 |
| Human | hsa-let-7a-5p | MIMAT0000062 |
Stem-loop Details
Human

Genomic Map